siRNA Details
| Gene Symbol: 4921517D16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK040269.1 | - | 2 | 517 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB635723.1 | - | 2 | 515 | Chr. 16 - 57338311 - 57339977 [+] on Build GRCm38 | |

4921517D16Rik
Genomic Map