siRNA Details
| Gene Symbol: LOC102632465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131873.1 | - | 3 | 719 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102632465| Gene Symbol: LOC102632465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131873.1 | - | 3 | 719 | ||