siRNA Details
| Gene Symbol: LOC102632465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK131873.1 | - | 3 | 671 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY708442.1 | - | 3 | 664 | Chr. 2 - 6472000 - 6478498 [+] on Build GRCm38 | |

LOC102632465
Genomic Map