siRNA Details
| Gene Symbol: Gm17830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033486.1 | 5 | 1657 | |||
| GenBank mRNA | AK135726.1 | - | 5 | 1658 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm17830| Gene Symbol: Gm17830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033486.1 | 5 | 1657 | |||
| GenBank mRNA | AK135726.1 | - | 5 | 1658 | ||