siRNA Details
| Gene Symbol: Gm5122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040767.1 | 3 | 1279 | |||
| GenBank mRNA | AK040484.1 | - | 3 | 1279 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5122| Gene Symbol: Gm5122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040767.1 | 3 | 1279 | |||
| GenBank mRNA | AK040484.1 | - | 3 | 1279 | ||