siRNA Details
| Gene Symbol: Gm16087 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132125.1 | - | 1 | 358 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY713232.1 | - | 1 | 354 | Chr. 12 - 106954767 - 106955924 [-] on Build GRCm38 | |

Gm16087
Genomic Map