siRNA Details
| Gene Symbol: Nsun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020172.1 | - | 1 | 312 | ||
| AK088718.1 | - | 1 | 312 | |||
| AK162988.1 | - | 1 | 302 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsun3| Gene Symbol: Nsun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020172.1 | - | 1 | 312 | ||
| AK088718.1 | - | 1 | 312 | |||
| AK162988.1 | - | 1 | 302 | |||