siRNA Details
| Gene Symbol: Nsun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020172.1 | - | 1 | 1255 | ||
| AK088718.1 | - | 1 | 1255 | |||
| AK162988.1 | - | 1 | 1246 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY762969.1 | - | 1 | 393 | Chr. 16 - 62785237 - 62785905 [-] on Build GRCm38 | |

Nsun3
Genomic Map