siRNA Details
| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046449.1 | - | 1 | 2374 | ||
| AK081215.1 | - | 1 | 1049 | |||
| BB283956.1 | - | 1 | 112 | |||
| BB355593.2 | - | 1 | 209 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbck| Gene Symbol: Tbck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046449.1 | - | 1 | 2374 | ||
| AK081215.1 | - | 1 | 1049 | |||
| BB283956.1 | - | 1 | 112 | |||
| BB355593.2 | - | 1 | 209 | |||