siRNA Details
| Gene Symbol: A330076C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045088.1 | 4 | 1736 | |||
| GenBank mRNA | AK039489.1 | - | 4 | 1737 | ||
| AK039634.1 | - | 4 | 1736 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330076C08Rik| Gene Symbol: A330076C08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045088.1 | 4 | 1736 | |||
| GenBank mRNA | AK039489.1 | - | 4 | 1737 | ||
| AK039634.1 | - | 4 | 1736 | |||