siRNA Details
Gene Symbol: A330076C08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045088.1 | 4 | 1909 | |||
GenBank mRNA | AK039489.1 | - | 4 | 1910 | ||
AK039634.1 | - | 4 | 1909 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A330076C08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045088.1 | 4 | 1909 | |||
GenBank mRNA | AK039489.1 | - | 4 | 1910 | ||
AK039634.1 | - | 4 | 1909 |