siRNA Details
| Gene Symbol: Gm37359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144610.1 | - | 85 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY733981.1 | - | 2 | 85 | Chr. 3 - 65829116 - 65872281 [-] on Build GRCm38 | |

Gm37359
Genomic Map