siRNA Details
| Gene Symbol: 2900064B18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013747.1 | - | 1 | 1082 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE985970.1 | - | 1 | 311 | Chr. 16 - 23985615 - 23985940 [-] on Build GRCm38 | |

2900064B18Rik
Genomic Map