siRNA Details
| Gene Symbol: 4930474N09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038130.1 | 2 | 379 | |||
| GenBank mRNA | AK015577.1 | - | 2 | 282 | ||
| AK019719.1 | - | 2 | 269 | |||
| BY100105.1 | - | 2 | 379 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715126.1 | - | 2 | 282 | Chr. 12 - 99161426 - 99162709 [-] on Build GRCm38 | |

4930474N09Rik
Genomic Map