siRNA Details
| Gene Symbol: 4930474N09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038130.1 | 1, 2 | 259 | |||
| GenBank mRNA | AK015577.1 | - | 1, 2 | 162 | ||
| AK019719.1 | - | 1, 2 | 149 | |||
| BY100105.1 | - | 1, 2 | 259 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930474N09Rik| Gene Symbol: 4930474N09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038130.1 | 1, 2 | 259 | |||
| GenBank mRNA | AK015577.1 | - | 1, 2 | 162 | ||
| AK019719.1 | - | 1, 2 | 149 | |||
| BY100105.1 | - | 1, 2 | 259 | |||