siRNA Details
| Gene Symbol: 2900064F13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013749.1 | - | 1 | 349 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ261689.1 | - | 1 | 49 | Chr. 5 - 81827118 - 81827823 [+] on Build GRCm38 | |

2900064F13Rik
Genomic Map