siRNA Details
| Gene Symbol: 4930529L06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040537.1 | 1 | 66 | |||
| GenBank mRNA | AK019711.1 | - | 1 | 67 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715499.1 | - | 1 | 67 | Chr. 16 - 84679789 - 84685150 [+] on Build GRCm38 | |

4930529L06Rik
Genomic Map