siRNA Details
| Gene Symbol: 5830408C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017910.1 | - | 1 | 1155 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BF658642.1 | - | 1 | 254 | Chr. 8 - 25108945 - 25109467 [+] on Build GRCm38 | |

5830408C22Rik
Genomic Map