siRNA Details
| Gene Symbol: Slx4ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK158198.1 | - | 2 | 3061 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AK171727.1 | - | 3862 | Chr. - - [-] on Build | ||

Slx4ip
Genomic Map