siRNA Details
| Gene Symbol: C330006D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021182.1 | - | 1 | 735 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV050805.1 | - | 1 | 563 | Chr. 9 - 7836009 - 7836862 [-] on Build GRCm38 | |

C330006D17Rik
Genomic Map