siRNA Details
| Gene Symbol: Gm16348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050994.1 | - | 2 | 2176 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16348| Gene Symbol: Gm16348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050994.1 | - | 2 | 2176 | ||