siRNA Details
| Gene Symbol: Gm16348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050994.1 | - | 2 | 1798 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DV046811.1 | - | 3 | 758 | Chr. 8 - 41218140 - 41220515 [+] on Build GRCm38 | |

Gm16348
Genomic Map