siRNA Details
| Gene Symbol: Gm13446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045894.1 | 4 | 1867 | |||
| GenBank mRNA | AK138201.1 | - | 4 | 1868 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13446| Gene Symbol: Gm13446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045894.1 | 4 | 1867 | |||
| GenBank mRNA | AK138201.1 | - | 4 | 1868 | ||