siRNA Details
| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013969.1 | - | 1 | 434 | ||
| AK013971.1 | - | 1 | 434 | |||
| AK170049.1 | - | 1 | 626 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex261| Gene Symbol: Tex261 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013969.1 | - | 1 | 434 | ||
| AK013971.1 | - | 1 | 434 | |||
| AK170049.1 | - | 1 | 626 | |||