siRNA Details
Gene Symbol: 3110001N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013968.1 | - | 1 | 139 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712984.1 | - | 1 | 136 | Chr. 5 - 131525003 - 131525660 [-] on Build GRCm38 |
