siRNA Details
| Gene Symbol: Slc30a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020581.1 | - | 1 | 486 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722072.1 | - | 1 | 483 | Chr. 17 - 74395641 - 74396199 [+] on Build GRCm38 | |

Slc30a6
Genomic Map