siRNA Details
| Gene Symbol: 1600002D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040484.1 | 4 | 384 | |||
| GenBank mRNA | AK005393.1 | - | 4 | 385 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705598.1 | - | 4 | 382 | Chr. 16 - 95841886 - 95929075 [-] on Build GRCm38 | |

1600002D24Rik
Genomic Map