siRNA Details
| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK042235.1 | - | 2 | 578 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB238783.2 | - | 2 | 588 | Chr. 3 - 33131571 - 33143088 [-] on Build GRCm38 | |

Pex5l
Genomic Map