siRNA Details
| Gene Symbol: 1700019F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006117.1 | - | 2 | 377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700019F05Rik| Gene Symbol: 1700019F05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006117.1 | - | 2 | 377 | ||