siRNA Details
| Gene Symbol: 1700003L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040507.1 | 1, 2 | 138 | |||
| GenBank mRNA | AK005649.1 | - | 1, 2 | 139 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700003L19Rik| Gene Symbol: 1700003L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040507.1 | 1, 2 | 138 | |||
| GenBank mRNA | AK005649.1 | - | 1, 2 | 139 | ||