siRNA Details
| Gene Symbol: 1700003K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005648.1 | - | 1 | 203 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705839.1 | - | 1 | 203 | Chr. 14 - 71946794 - 71947213 [+] on Build GRCm38 | |

1700003K11Rik
Genomic Map