siRNA Details
| Gene Symbol: Gm11837 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005641.1 | - | 6 | 576 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ189634.1 | - | 2 | 369 | Chr. 4 - 14947479 - 14953026 [+] on Build GRCm38 | |

Gm11837
Genomic Map