siRNA Details
| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037908.1 | - | 4286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sptlc2| Gene Symbol: Sptlc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037908.1 | - | 4286 | |||