siRNA Details
| Gene Symbol: 5830408C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037533.1 | - | 1 | 2789 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB155240.2 | - | 510 | Chr. - - [-] on Build | ||

5830408C22Rik
Genomic Map