siRNA Details
| Gene Symbol: 1700020I14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004510.1 | - | 902 | |||
| AK147270.1 | - | 7737 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700020I14Rik| Gene Symbol: 1700020I14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK004510.1 | - | 902 | |||
| AK147270.1 | - | 7737 | ||||