siRNA Details
| Gene Symbol: 4930565N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040476.1 | 3 | 854 | |||
| GenBank mRNA | AK015360.1 | - | 3 | 857 | ||
| AK019787.1 | - | 3 | 804 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930565N06Rik| Gene Symbol: 4930565N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040476.1 | 3 | 854 | |||
| GenBank mRNA | AK015360.1 | - | 3 | 857 | ||
| AK019787.1 | - | 3 | 804 | |||