siRNA Details
Gene Symbol: 4930565N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040476.1 | 3 | 1064 | |||
GenBank mRNA | AK015360.1 | - | 3 | 1067 | ||
AK019787.1 | - | 3 | 1014 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930565N06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040476.1 | 3 | 1064 | |||
GenBank mRNA | AK015360.1 | - | 3 | 1067 | ||
AK019787.1 | - | 3 | 1014 |