siRNA Details
| Gene Symbol: 4930422M22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015183.1 | - | 2 | 712 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714734.1 | - | 2 | 713 | Chr. 9 - 99299038 - 99304675 [-] on Build GRCm38 | |

4930422M22Rik
Genomic Map