siRNA Details
| Gene Symbol: 5730510P18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017761.1 | - | 1 | 276 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF804828.1 | - | 1 | 297 | Chr. 12 - 12997172 - 12997651 [+] on Build GRCm38 | |

5730510P18Rik
Genomic Map