siRNA Details
| Gene Symbol: D5Ertd615e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043596.1 | - | 779 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D5Ertd615e| Gene Symbol: D5Ertd615e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043596.1 | - | 779 | |||