siRNA Details
| Gene Symbol: 4930535F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015981.1 | - | 1 | 659 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715551.1 | - | 1 | 657 | Chr. 19 - 48991019 - 48991969 [-] on Build GRCm38 | |

4930535F04Rik
Genomic Map