siRNA Details
| Gene Symbol: 4933430A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016987.1 | - | 2 | 1357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933430A20Rik| Gene Symbol: 4933430A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016987.1 | - | 2 | 1357 | ||