siRNA Details
| Gene Symbol: 4933429H19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016981.1 | - | 1 | 800 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB274652.1 | - | 505 | Chr. 16 - 30362660 - 30388530 [-] on Build GRCm38 | ||

4933429H19Rik
Genomic Map