siRNA Details
| Gene Symbol: Rap1gapos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007899.1 | - | 2 | 300 | ||
| BQ287036.1 | - | 1 | 148 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707902.1 | - | 2 | 300 | Chr. 4 - 137696977 - 137706606 [-] on Build GRCm38 | |

Rap1gapos
Genomic Map