siRNA Details
| Gene Symbol: 2900019A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013553.1 | - | 1 | 635 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV151516.1 | - | 1 | 131 | Chr. 5 - 24903368 - 24903656 [-] on Build GRCm38 | |

2900019A20Rik
Genomic Map