siRNA Details
| Gene Symbol: 2900018K06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK013551.1 | - | 1 | 229 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY712516.1 | - | 1 | 227 | Chr. 2 - 136120255 - 136120721 [-] on Build GRCm38 | |

2900018K06Rik
Genomic Map