siRNA Details
| Gene Symbol: 4930562A09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016187.1 | - | 1 | 442 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715795.1 | - | 1 | 442 | Chr. 5 - 116390678 - 116392794 [-] on Build GRCm38 | |

4930562A09Rik
Genomic Map