siRNA Details
| Gene Symbol: Fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038226.1 | - | 6 | 3094 | ||
| AK042244.1 | - | 1 | 75 | |||
| AK084867.1 | - | 1 | 808 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fus| Gene Symbol: Fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK038226.1 | - | 6 | 3094 | ||
| AK042244.1 | - | 1 | 75 | |||
| AK084867.1 | - | 1 | 808 | |||