siRNA Details
| Gene Symbol: Cdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029710.1 | - | 2503 | |||
| AK134896.1 | - | 1033 | ||||
| AK153645.1 | - | 1418 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc73| Gene Symbol: Cdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK029710.1 | - | 2503 | |||
| AK134896.1 | - | 1033 | ||||
| AK153645.1 | - | 1418 | ||||