siRNA Details
| Gene Symbol: Gm12758 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087850.1 | - | 674 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB235913.1 | - | 2 | 670 | Chr. 7 - 34234381 - 34236777 [+] on Build GRCm38 | |

Gm12758
Genomic Map